Assay Details
| Gene Symbol: LOC4324711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765311.2 | XP_015620797.1 | 7 - 8 | 973 | 72 | |
| XM_015765321.2 | 3 - 4 | 228 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4324711| Gene Symbol: LOC4324711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765311.2 | XP_015620797.1 | 7 - 8 | 973 | 72 | |
| XM_015765321.2 | 3 - 4 | 228 | 72 | |||
Set Membership: |
|