Assay Details
| Gene Symbol: LOC4324711 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765311.2 | XP_015620797.1 | 9 - 10 | 1088 | 97 | |
| XM_015765321.2 | 5 - 6 | 343 | 97 | |||
More Information
Set Membership: |
|

LOC4324711
Genomic Map