Assay Details
| Gene Symbol: LOC4327367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765764.2 | XP_015621250.1 | 6 - 7 | 1122 | 97 | |
| GenBank mRNA | AB079063.2 | - | 6 - 7 | 1044 | 97 | |
| AK069181.1 | - | 5 - 6 | 1862 | 97 | ||
| AK104675.1 | - | 4 - 5 | 285 | 97 | ||
| BT129083.1 | - | 3 - 4 | 246 | 97 | ||
| BT129117.1 | - | 3 - 4 | 245 | 97 | ||
More Information
Set Membership: |
|

LOC4327367
Genomic Map