Assay Details
| Gene Symbol: LOC4327497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015761030.2 | XP_015616516.1 | 2 - 3 | 302 | 58 | |
| GenBank mRNA | AK243099.1 | - | 2 - 3 | 172 | 58 | |
More Information
Set Membership: |
|

LOC4327497
Genomic Map