Assay Details
| Gene Symbol: LOC4325072 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785997.2 | XP_015641483.1 | 3 - 4 | 350 | 72 | |
| GenBank mRNA | AK111757.1 | - | 3 - 4 | 296 | 72 | |
More Information
Set Membership: |
|

LOC4325072
Genomic Map