Assay Details
| Gene Symbol: LOC4325084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765735.2 | XP_015621221.1 | 3 - 4 | 457 | 119 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4325084| Gene Symbol: LOC4325084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765735.2 | XP_015621221.1 | 3 - 4 | 457 | 119 | |
Set Membership: |
|