Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 10 - 11 | 1816 | 80 | |
| GenBank mRNA | AK067525.1 | - | 8 - 9 | 1329 | 80 | |
| AK070047.1 | - | 10 - 11 | 1767 | 80 | ||
More Information
Set Membership: |
|

LOC4325748
Genomic Map