Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 13 - 14 | 2217 | 144 | |
| GenBank mRNA | AK067525.1 | - | 11 - 12 | 1730 | 144 | |
| AK070047.1 | - | 13 - 14 | 2168 | 144 | ||
More Information
Set Membership: |
|

LOC4325748
Genomic Map