Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 16 - 17 | 2698 | 88 | |
| GenBank mRNA | AK067525.1 | - | 14 - 15 | 2211 | 88 | |
| AK070047.1 | - | 16 - 17 | 2648 | 88 | ||
More Information
Set Membership: |
|

LOC4325748
Genomic Map