Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 2 - 3 | 354 | 56 | |
| GenBank mRNA | AK070047.1 | - | 2 - 3 | 305 | 56 | |
More Information
Set Membership: |
|

LOC4325748
Genomic Map