Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 7 - 8 | 1066 | 65 | |
| GenBank mRNA | AK067525.1 | - | 5 - 6 | 579 | 65 | |
| AK070047.1 | - | 7 - 8 | 1017 | 65 | ||
More Information
Set Membership: |
|

LOC4325748
Genomic Map