Assay Details
| Gene Symbol: LOC4325748 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764568.2 | XP_015620054.1 | 9 - 10 | 1505 | 122 | |
| GenBank mRNA | AK067525.1 | - | 7 - 8 | 1018 | 122 | |
| AK070047.1 | - | 9 - 10 | 1456 | 122 | ||
More Information
Set Membership: |
|

LOC4325748
Genomic Map