Assay Details
| Gene Symbol: LOC4328021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767729.2 | XP_015623215.1 | 4 - 5 | 1342 | 100 | |
| XM_015767730.2 | XP_015623216.1 | 4 - 5 | 1336 | 100 | ||
| XM_015767731.2 | XP_015623217.1 | 4 - 5 | 1336 | 100 | ||
| GenBank mRNA | AK073056.1 | - | 4 - 5 | 1343 | 100 | |
More Information
Set Membership: |
|

LOC4328021
Genomic Map