Assay Details
| Gene Symbol: LOC4328060 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770786.1 | XP_015626272.1 | 7 - 8 | 1276 | 110 | |
| GenBank mRNA | AK061078.1 | - | 7 - 8 | 1241 | 110 | |
| AK073508.1 | - | 7 - 8 | 1242 | 110 | ||
| AK103982.1 | - | 1 - 2 | 112 | 110 | ||
More Information
Set Membership: |
|

LOC4328060
Genomic Map