Assay Details
| Gene Symbol: LOC9271200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768016.2 | XP_015623502.1 | 5 - 6 | 639 | 65 | |
| XM_015768019.2 | XP_015623505.1 | 5 - 6 | 639 | 65 | ||
| XM_015768020.2 | XP_015623506.1 | 5 - 6 | 639 | 65 | ||
| GenBank mRNA | AK073966.1 | - | 3 - 4 | 1551 | 65 | |
More Information
Set Membership: |
|

LOC9271200
Genomic Map