Assay Details
| Gene Symbol: LOC9272150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771957.2 | XP_015627443.1 | 4 - 5 | 482 | 88 | |
| XM_015771958.2 | XP_015627444.1 | 4 - 5 | 482 | 88 | ||
| XM_015771959.2 | XP_015627445.1 | 4 - 5 | 396 | 88 | ||
| XM_015771960.2 | XP_015627446.1 | 4 - 5 | 482 | 88 | ||
| GenBank mRNA | AK073717.1 | - | 4 - 5 | 397 | 88 | |
More Information
Set Membership: |
|

LOC9272150
Genomic Map