Assay Details
| Gene Symbol: LOC9272150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771957.2 | XP_015627443.1 | 6 - 7 | 635 | 128 | |
| XM_015771958.2 | XP_015627444.1 | 6 - 7 | 635 | 128 | ||
| XM_015771959.2 | XP_015627445.1 | 6 - 7 | 549 | 128 | ||
| XM_015771960.2 | XP_015627446.1 | 6 - 7 | 635 | 128 | ||
| GenBank mRNA | AK073717.1 | - | 6 - 7 | 550 | 128 | |
More Information
Set Membership: |
|

LOC9272150
Genomic Map