Assay Details
| Gene Symbol: LOC4346173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795284.2 | XP_015650770.1 | 10 - 11 | 2001 | 126 | |
| XM_015795285.2 | XP_015650771.1 | 10 - 11 | 1999 | 126 | ||
| GenBank mRNA | AK111766.1 | - | 10 - 11 | 1770 | 126 | |
| AK112001.1 | - | 10 - 11 | 1466 | 126 | ||
More Information
Set Membership: |
|

LOC4346173
Genomic Map