Assay Details
| Gene Symbol: LOC4346173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795284.2 | XP_015650770.1 | 7 - 8 | 1663 | 114 | |
| XM_015795285.2 | XP_015650771.1 | 7 - 8 | 1661 | 114 | ||
| GenBank mRNA | AK111766.1 | - | 7 - 8 | 1432 | 114 | |
| AK112001.1 | - | 7 - 8 | 1128 | 114 | ||
More Information
Set Membership: |
|

LOC4346173
Genomic Map