Assay Details
| Gene Symbol: LOC4346176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793186.2 | XP_015648672.1 | 10 - 11 | 2219 | 71 | |
| XM_015793187.2 | XP_015648673.1 | 10 - 11 | 2219 | 71 | ||
| GenBank mRNA | AK240830.1 | - | 10 - 11 | 2141 | 71 | |
| AK241627.1 | - | 10 - 11 | 2182 | 71 | ||
| AY941774.1 | - | 10 - 11 | 2096 | 71 | ||
More Information
Set Membership: |
|

LOC4346176
Genomic Map