Assay Details
| Gene Symbol: LOC4346176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793186.2 | XP_015648672.1 | 12 - 13 | 2436 | 107 | |
| XM_015793187.2 | XP_015648673.1 | 12 - 13 | 2436 | 107 | ||
| GenBank mRNA | AK240830.1 | - | 12 - 13 | 2358 | 107 | |
| AK241627.1 | - | 12 - 13 | 2399 | 107 | ||
| AY941774.1 | - | 12 - 13 | 2313 | 107 | ||
More Information
Set Membership: |
|

LOC4346176
Genomic Map