Assay Details
| Gene Symbol: LOC4346176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793186.2 | XP_015648672.1 | 17 - 18 | 3100 | 101 | |
| XM_015793187.2 | XP_015648673.1 | 17 - 18 | 3097 | 101 | ||
| GenBank mRNA | AK240830.1 | - | 17 - 18 | 3019 | 101 | |
| AK241627.1 | - | 17 - 18 | 3060 | 101 | ||
| AY941774.1 | - | 17 - 18 | 2974 | 101 | ||
More Information
Set Membership: |
|

LOC4346176
Genomic Map