Assay Details
| Gene Symbol: LOC4346176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793186.2 | XP_015648672.1 | 9 - 10 | 1348 | 63 | |
| XM_015793187.2 | XP_015648673.1 | 9 - 10 | 1348 | 63 | ||
| GenBank mRNA | AK240830.1 | - | 9 - 10 | 1270 | 63 | |
| AK241627.1 | - | 9 - 10 | 1311 | 63 | ||
| AY941774.1 | - | 9 - 10 | 1225 | 63 | ||
More Information
Set Membership: |
|

LOC4346176
Genomic Map