Assay Details
Gene Symbol: LOC4346180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795164.2 | XP_015650650.1 | 2 - 3 | 456 | 84 | |
GenBank mRNA | AK067119.1 | - | 2 - 3 | 441 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
