Assay Details
| Gene Symbol: LOC4346211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794438.2 | XP_015649924.1 | 1 - 2 | 190 | 98 | |
| GenBank mRNA | AK071354.1 | - | 1 - 2 | 79 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794438.2 transcripts.
Set Membership: |
|

LOC4346211
Genomic Map