Assay Details
| Gene Symbol: LOC4346211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794438.2 | XP_015649924.1 | 22 - 23 | 3590 | 100 | |
| GenBank mRNA | AK059291.1 | - | 1 - 2 | 705 | 100 | |
| AK071354.1 | - | 22 - 23 | 3480 | 100 | ||
More Information
Set Membership: |
|

LOC4346211
Genomic Map