Assay Details
| Gene Symbol: LOC4346211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794438.2 | XP_015649924.1 | 4 - 5 | 730 | 115 | |
| GenBank mRNA | AK071354.1 | - | 4 - 5 | 620 | 115 | |
More Information
Set Membership: |
|

LOC4346211
Genomic Map