Assay Details
| Gene Symbol: LOC4351589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764803.2 | XP_015620289.1 | 4 - 5 | 961 | 95 | |
| XM_015764805.2 | XP_015620291.1 | 3 - 4 | 310 | 95 | ||
| GenBank mRNA | AK242705.1 | - | 4 - 5 | 962 | 95 | |
More Information
Set Membership: |
|

LOC4351589
Genomic Map