Assay Details
| Gene Symbol: LOC4352275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764379.2 | 4 - 5 | 440 | 138 | ||
| GenBank mRNA | AY250075.1 | - | 4 - 5 | 440 | 138 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352275| Gene Symbol: LOC4352275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764379.2 | 4 - 5 | 440 | 138 | ||
| GenBank mRNA | AY250075.1 | - | 4 - 5 | 440 | 138 | |
Set Membership: |
|