Assay Details
| Gene Symbol: LOC4352275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764379.2 | 6 - 7 | 592 | 68 | ||
| GenBank mRNA | AY250075.1 | - | 6 - 7 | 592 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352275| Gene Symbol: LOC4352275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764379.2 | 6 - 7 | 592 | 68 | ||
| GenBank mRNA | AY250075.1 | - | 6 - 7 | 592 | 68 | |
Set Membership: |
|