Assay Details
| Gene Symbol: LOC4352306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765120.2 | XP_015620606.1 | 2 - 3 | 646 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352306| Gene Symbol: LOC4352306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765120.2 | XP_015620606.1 | 2 - 3 | 646 | 100 | |
Set Membership: |
|