Assay Details
| Gene Symbol: LOC4352341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762686.2 | XP_015618172.1 | 4 - 5 | 546 | 76 | |
| GenBank mRNA | AK101966.1 | - | 4 - 5 | 535 | 76 | |
More Information
Set Membership: |
|

LOC4352341
Genomic Map