Assay Details
| Gene Symbol: LOC4352420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764693.2 | XP_015620179.1 | 4 - 5 | 1079 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4352420| Gene Symbol: LOC4352420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764693.2 | XP_015620179.1 | 4 - 5 | 1079 | 120 | |
Set Membership: |
|