Assay Details
| Gene Symbol: LOC4352514 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763342.2 | XP_015618828.1 | 5 - 6 | 664 | 109 | |
| GenBank mRNA | AK073411.1 | - | 5 - 6 | 671 | 109 | |
More Information
Set Membership: |
|

LOC4352514
Genomic Map