Assay Details
| Gene Symbol: LOC4352706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764575.1 | XP_015620061.1 | 8 - 9 | 1043 | 89 | |
| GenBank mRNA | AK101904.1 | - | 8 - 9 | 1050 | 89 | |
| AK103799.1 | - | 8 - 9 | 815 | 89 | ||
More Information
Set Membership: |
|

LOC4352706
Genomic Map