Assay Details
| Gene Symbol: LOC4352850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765050.2 | XP_015620536.1 | 3 - 4 | 337 | 81 | |
| XM_015765051.2 | XP_015620537.1 | 3 - 4 | 357 | 81 | ||
| GenBank mRNA | AK061110.1 | - | 3 - 4 | 327 | 81 | |
| AK069213.1 | - | 3 - 4 | 305 | 81 | ||
More Information
Set Membership: |
|

LOC4352850
Genomic Map