Assay Details
| Gene Symbol: LOC4352850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765050.2 | XP_015620536.1 | 4 - 5 | 538 | 86 | |
| XM_015765051.2 | XP_015620537.1 | 4 - 5 | 558 | 86 | ||
| GenBank mRNA | AK061110.1 | - | 4 - 5 | 528 | 86 | |
| AK069213.1 | - | 4 - 5 | 506 | 86 | ||
More Information
Set Membership: |
|

LOC4352850
Genomic Map