Assay Details
| Gene Symbol: LOC4350611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759867.2 | XP_015615353.1 | 2 - 3 | 378 | 68 | |
| GenBank mRNA | AK106093.1 | - | 2 - 3 | 333 | 68 | |
More Information
Set Membership: |
|

LOC4350611
Genomic Map