Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 13 - 14 | 2646 | 71 | |
| GenBank mRNA | AK099809.1 | - | 6 - 7 | 1001 | 71 | |
| AK120821.1 | - | 10 - 11 | 1842 | 71 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map