Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 18 - 19 | 3194 | 145 | |
| GenBank mRNA | AK099809.1 | - | 11 - 12 | 1549 | 145 | |
| AK120821.1 | - | 15 - 16 | 2389 | 145 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map