Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 22 - 23 | 3886 | 79 | |
| GenBank mRNA | AK099809.1 | - | 15 - 16 | 2241 | 79 | |
| AK120821.1 | - | 19 - 20 | 3081 | 79 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map