Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 5 - 6 | 1060 | 72 | |
| GenBank mRNA | AK120821.1 | - | 2 - 3 | 256 | 72 | |
More Information
Set Membership: |
|

LOC4337574
Genomic Map