Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 9 - 10 | 1972 | 97 | |
| GenBank mRNA | AK099809.1 | - | 2 - 3 | 327 | 97 | |
| AK120821.1 | - | 6 - 7 | 1168 | 97 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map