Assay Details
| Gene Symbol: LOC4337574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784553.2 | XP_015640039.1 | 12 - 13 | 2548 | 64 | |
| GenBank mRNA | AK099809.1 | - | 5 - 6 | 903 | 64 | |
| AK120821.1 | - | 9 - 10 | 1744 | 64 | ||
More Information
Set Membership: |
|

LOC4337574
Genomic Map