Assay Details
| Gene Symbol: LOC4338469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782254.2 | XP_015637740.1 | 1 - 2 | 249 | 69 | |
| GenBank mRNA | AK111385.1 | - | 1 - 2 | 194 | 69 | |
More Information
Set Membership: |
|

LOC4338469
Genomic Map