Assay Details
| Gene Symbol: LOC4338480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784840.1 | XP_015640326.1 | 17 - 18 | 2229 | 113 | |
| XM_015784841.2 | XP_015640327.1 | 15 - 16 | 1811 | 113 | ||
| GenBank mRNA | AK073748.1 | - | 17 - 18 | 2224 | 113 | |
| AK102482.1 | - | 17 - 18 | 2223 | 113 | ||
More Information
Set Membership: |
|

LOC4338480
Genomic Map