Assay Details
| Gene Symbol: LOC9267485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782797.2 | XP_015638283.1 | 11 - 12 | 1100 | 148 | |
| XM_015782799.2 | XP_015638285.1 | 11 - 12 | 1080 | 148 | ||
| GenBank mRNA | AK073973.1 | - | 7 - 8 | 1788 | 148 | |
More Information
Set Membership: |
|

LOC9267485
Genomic Map