Assay Details
| Gene Symbol: LOC4339576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781987.2 | XP_015637473.1 | 1 - 2 | 443 | 95 | |
| GenBank mRNA | AK069566.1 | - | 1 - 2 | 311 | 95 | |
More Information
Set Membership: |
|

LOC4339576
Genomic Map