Assay Details
| Gene Symbol: LOC4339576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781987.2 | XP_015637473.1 | 6 - 7 | 1311 | 67 | |
| GenBank mRNA | AK069566.1 | - | 6 - 7 | 1179 | 67 | |
More Information
Set Membership: |
|

LOC4339576
Genomic Map