Assay Details
| Gene Symbol: LOC4340564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788359.2 | XP_015643845.1 | 2 - 3 | 1692 | 84 | |
| GenBank mRNA | AK070606.1 | - | 2 - 3 | 1489 | 84 | |
More Information
Set Membership: |
|

LOC4340564
Genomic Map